MilliporeSigma
  • Down-regulation of pyrophosphate: fructose 6-phosphate 1-phosphotransferase (PFP) activity in sugarcane enhances sucrose accumulation in immature internodes.
Down-regulation of pyrophosphate: fructose 6-phosphate 1-phosphotransferase (PFP) activity in sugarcane enhances sucrose accumulation in immature internodes.
Transgenic research (2007-04-14)
Jan-Hendrik Groenewald, Frederik Coenraad Botha
ABSTRACT
Pyrophosphate: fructose 6-phosphate 1-phosphotransferase (PFP) activity was successfully down-regulated in sugarcane using constitutively expressed antisense and untranslatable forms of the sugarcane PFP-beta gene. In young internodal tissue activity was reduced by up to 70% while no residual activity could be detected in mature tissues. The transgenic plants showed no visible phenotype or significant differences in growth and development under greenhouse and field conditions. Sucrose concentrations were significantly increased in the immature internodes of the transgenic plants but not in the mature internodes. This contributed to an increase in the purity of the immature tissues, resembling an early ripening phenotype. Both the immature and mature internodes of the transgenic plants had significantly higher fibre contents. These findings suggest that PFP influences the ability of young, biosynthetically active sugarcane culm tissue to accumulate sucrose but that the equilibrium of the glycolytic intermediates, including the stored sucrose, is restored when ATP-dependent phosphofructokinase and the residual PFP activity is sufficient to sustain the required glycolytic flux as the tissue matures. Moreover, it suggests a role for PFP in glycolytic carbon flow, which could be rate limiting under conditions of high metabolic activity.