MilliporeSigma
  • Functional expression of porcine aminoacylase 1 in E. coli using a codon optimized synthetic gene and molecular chaperones.
Functional expression of porcine aminoacylase 1 in E. coli using a codon optimized synthetic gene and molecular chaperones.
Applied microbiology and biotechnology (2008-09-26)
Rainer Wardenga, Frank Hollmann, Oliver Thum, Uwe Bornscheuer
ABSTRACT
Efficient recombinant expression of N-acyl-L-aminoacylase 1 from pig kidney (pAcy1) was achieved in the prokaryotic host Escherichia coli. An optimized nucleotide sequence (codon adaptation index 0.95 for E. coli), was cloned into vector pET-52(b) yielding an E. coli-expressible pAcy1 gene. Formation of inclusion bodies was alleviated by co-expression of molecular chaperones resulting in 2.7- and 4.2-fold increased recovery of active pAcy1 using trigger factor or GroEL-GroES, respectively. Facile purification was achieved via StrepTag affinity chromatography. Overall, more than 80 mg highly active pAcy1 (94 U/mg) was obtained per liter of cultivation broth. The protein was analyzed for structural and functional identity, and the performances of further described expression and purification systems for pAcy1 were compared.
Sigma-Aldrich
Acylase I from Aspergillus melleus, powder, brown, >0.5 U/mg
Sigma-Aldrich
Acylase I from porcine kidney, Grade I, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Acylase I from porcine kidney, Grade II, salt-free, lyophilized powder, 300-1,500 units/mg protein