MilliporeSigma
Thermus thermophilus strains active in purine nucleoside synthesis.
Molecules (Basel, Switzerland) (2009-03-28)
Marcos Almendros, José-Vicente Sinisterra Gago, José Berenguer Carlos
ABSTRACT
Several strains of Thermus thermophilus were tested in order to detect purine nucleoside synthase activity using pyrimidine nucleosides as the sugar-donor and adenine or hypoxanthine as bases. High productivity values (t =1 hr) were obtained while completely avoiding adenosine-deaminase degradation of the products. N-2-deoxy-ribosyltransferase activity is described for the first time in hyperthermophilic bacteria.