MilliporeSigma
  • ApuA, a multifunctional alpha-glucan-degrading enzyme of Streptococcus suis, mediates adhesion to porcine epithelium and mucus.
ApuA, a multifunctional alpha-glucan-degrading enzyme of Streptococcus suis, mediates adhesion to porcine epithelium and mucus.
Microbiology (Reading, England) (2010-06-05)
Maria Laura Ferrando, Susana Fuentes, Astrid de Greeff, Hilde Smith, Jerry M Wells
ABSTRACT
We have identified apuA in Streptococcus suis, which encodes a bifunctional amylopullulanase with conserved alpha-amylase and pullulanase substrate-binding domains and catalytic motifs. ApuA exhibited properties typical of a Gram-positive surface protein, with a putative signal sequence and LPKTGE cell-wall-anchoring motif. A recombinant protein containing the predicted N-terminal alpha-amylase domain of ApuA was shown to have alpha-(1,4) glycosidic activity. Additionally, an apuA mutant of S. suis lacked the pullulanase alpha-(1,6) glycosidic activity detected in a cell-surface protein extract of wild-type S. suis. ApuA was required for normal growth in complex medium containing pullulan as the major carbon source, suggesting that this enzyme plays a role in nutrient acquisition in vivo via the degradation of glycogen and food-derived starch in the nasopharyngeal and oral cavities. ApuA was shown to promote adhesion to porcine epithelium and mucus in vitro, highlighting a link between carbohydrate utilization and the ability of S. suis to colonize and infect the host.
Sigma-Aldrich
Pullulanase microbial