MilliporeSigma
  • Purification of L-alpha glycerylphosphorylcholine by column chromatography.
Purification of L-alpha glycerylphosphorylcholine by column chromatography.
Journal of chromatography. A (2011-12-27)
Kangyi Zhang, Xingguo Wang, Jianhua Huang, Yuanfa Liu
ABSTRACT
Colorless L-alpha glycerylphosphorylcholine (L-α-GPC) was obtained at 99.8% purity, 69.8% recovery, and with a specific rotation of -2.5° via a five-step procedure. L-α-GPC was first produced by phospholipase A(1) hydrolysis of soy lecithin powder. Ca(2+) and Cl(-) were then effectively removed using two successive 001×7 cation and D311 anion exchange resin column chromatography procedures. Silica gel column chromatography and decoloration with active carbon were then applied to remove remaining impurities and colorant. Characterization of the L-α-GPC product was well in agreement with the standard. The resin and silica gel showed remarkable ability for L-α-GPC isolation after 10 uses. Thus, this study presents a simple and cost-effective method for preparing L-α-GPC with high yield and purity, low cost, and environmental friendliness, and encourages future investigation into its adaptation for industrial applications.
Sigma-Aldrich
Phospholipase A1 from Aspergillus oryzae