MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of 4,4'-dichloro-diphenyl diselenide (ClPhSe)2 on toxicity induced by mercuric chloride in mice: a comparative study with diphenyl diselenide (PhSe)2.
Effects of 4,4'-dichloro-diphenyl diselenide (ClPhSe)2 on toxicity induced by mercuric chloride in mice: a comparative study with diphenyl diselenide (PhSe)2.
Environmental toxicology and pharmacology (2012-09-18)
Mayara L de Freitas, Andreia R H da Silva, Silvane S Roman, Ricardo Brandão
ABSTRACT
The effects of 4,4'-dichloro-diphenyl diselenide (ClPhSe)(2) on the toxicity induced by mercuric chloride (HgCl(2)) were investigated and compared with diphenyl diselenide (PhSe)(2). Mice received HgCl(2) for three days and, on the third day, received (PhSe)(2) or (ClPhSe)(2). The results verified that the administration of (ClPhSe)(2) in mice exposed to HgCl(2) increased renal δ-aminolevulinate dehydratase (δ-ALA-D), Na(+), K(+)-ATPase activities and non-protein thiol (NPSH) levels and also decreased thiobarbituric acid-reactive substances (TBARS) and ascorbic acid levels, when compared to mice exposed to HgCl(2)+(PhSe)(2). Plasma and urinary protein, hemoglobin and hematocrit levels and histological parameters were also ameliorated in mice exposed to HgCl(2)+(ClPhSe)(2). In addition, the hepatic damage in mice exposed to HgCl(2)+(PhSe)(2) was reduced in animals exposed to (ClPhSe)(2). To sum up, the introduction of a functional group (chloro) in the aromatic ring of diaryl diselenide reduced the toxicity of this compound in liver and kidney of mice exposed to HgCl(2).
Sigma-Aldrich
Mercury standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Hg, 1000 ppm Hg
Sigma-Aldrich
Mercury(II) chloride, ≥98%
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)
Sigma-Aldrich
Diphenyl diselenide, 98%
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%