MilliporeSigma
  • Carbohydrate moieties of peanut agglutinin receptors isolated from human gastric cancer cells.
Carbohydrate moieties of peanut agglutinin receptors isolated from human gastric cancer cells.
Journal of biochemistry (1987-09-01)
N Shimoda, H Muramatsu, T Osawa, T Yamada, M Adachi, T Muramatsu
ABSTRACT
Receptors for peanut agglutinin (PNA) were isolated from Kato III human gastric cancer cells by affinity chromatography on PNA agarose, and were labeled by the galactose oxidase-NaB3H4 method. Alkaline NaBH4 treatment of the labeled receptors released two small oligosaccharide alcohols, which were identified as Gal beta 1----3GalNAc-ol and Gal beta 1----4GlcNAc beta 1----6(Gal beta 1----3)GalNAc-ol. Higher oligosaccharides and glycopeptides of both N- and O-linked type were also detected, but they did not appear to bear PNA binding sites. The presence of oligo-N-acetyllactosamine units in the N-linked type sugars was indicated by endo-beta-galactosidase digestion.