MilliporeSigma
  • ADAR1 protein induces adenosine-targeted DNA mutations in senescent Bcl6 gene-deficient cells.
ADAR1 protein induces adenosine-targeted DNA mutations in senescent Bcl6 gene-deficient cells.
The Journal of biological chemistry (2012-12-05)
Nobuhide Tsuruoka, Masafumi Arima, Nobuya Yoshida, Seiji Okada, Akemi Sakamoto, Masahiko Hatano, Hisae Satake, Eggi Arguni, Ji-Yang Wang, Jing-Hua Yang, Kazuko Nishikura, Souei Sekiya, Makio Shozu, Takeshi Tokuhisa
ABSTRACT
Somatic mutations accumulate in senescent cells. Bcl6, which functions as a transcriptional repressor, has been identified as a potent inhibitor of cell senescence, but a role of Bcl6 in the accumulation of somatic mutations has remained unclear. Ig class-switch recombination simultaneously induces somatic mutations in an IgM class-switch (Ig-Sμ) region of IgG B cells. Surprisingly, mutations were detected in the Ig-Sμ region of Bcl6-deficient IgM B cells without class-switch recombination, and these mutations were mainly generated by conversion of adenosine to guanosine, suggesting a novel DNA mutator in the B cells. The ADAR1 (adenosine deaminase acting on RNA1) gene was overexpressed in Bcl6-deficient cells, and its promoter analysis revealed that ADAR1 is a molecular target of Bcl6. Exogenous ADAR1 induced adenosine-targeted DNA mutations in IgM B cells from ADAR1-transgenic mice and in wild-type mouse embryonic fibroblasts (MEFs). These mutations accumulated in senescent MEFs accompanied with endogenous ADAR1 expression, and the frequency in senescent Bcl6-deficient MEFs was higher than senescent wild-type MEFs. Thus, Bcl6 protects senescent cells from accumulation of adenosine-targeted DNA mutations induced by ADAR1.
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type X, buffered aqueous glycerol solution, ≥130 units/mg protein
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type IX, ammonium sulfate suspension, 150-200 units/mg protein