MilliporeSigma
  • Influence of controlled pH on the activity of UDPG-pyrophosphorylase in Aureobasidium pullulans.
Influence of controlled pH on the activity of UDPG-pyrophosphorylase in Aureobasidium pullulans.
Carbohydrate polymers (2012-12-12)
Saikun Pan, Dongrui Yao, Jing Chen, Shengjun Wu
ABSTRACT
UDPG-pyrophosphorylase is the key enzyme involved in pullulan biosynthesis and pullulan production by Aureobasidium pullulans. In this study, effect of controlled pH on fermentation time, pullulan production, biomass, and UDPG-pyrophosphorylase activity was investigated. Pullulan yield increased to reach a maximum within 4 days, and maximum UDPG-pyrophosphorylase activity was observed at day 3, while the biomass continued to increase until the end of the experimental period. The A. pullulans isolated from sea mud grew well at relatively low pH. UDPG-pyrophosphorylase activity was affected by the controlled pH and reached a maximum at pH 5.5. Results indicated that UDPG-pyrophosphorylase activity was highly correlated with controlled pH and pullulan biosynthesis rate.
Product Description
Supelco
Pullulan Standard Set Mp ~350 - 700′000, set of analytical standards, suitable for gel permeation chromatography (GPC)
Sigma-Aldrich
Pullulan, suitable for substrate for pullulanase
Sigma-Aldrich
Uridine-5′-diphosphoglucose pyrophosphorylase from baker′s yeast, Type X, lyophilized powder, ≥40 units/mg protein