MilliporeSigma
Silver staining of native and denatured eucaryotic DNA in agarose gels.
Analytical biochemistry (1987-08-15)
M Gottlieb, M Chavko
ABSTRACT
A modified method of silver staining for native and denatured eucaryotic DNA in 1% agarose gel is described. This method is at least fivefold more sensitive than ethidium bromide staining, with a detection limit of 2.5 ng for total DNA. The calibration curve is linear within the range 5-30 ng of single-stranded and double-stranded DNA. This method is especially advantageous for electrophoretic assessment of DNA molecular weights.