MilliporeSigma
  • Isoelectric focusing in layers of granulated gels. I. Thin-layer isoelectric focusing of proteins.
Isoelectric focusing in layers of granulated gels. I. Thin-layer isoelectric focusing of proteins.
Biochimica et biophysica acta (1973-02-21)
B J Radola