MilliporeSigma
  • Evaluation of DA-9801, a new herbal drug for diabetic neuropathy, on metabolism-mediated interaction.
Evaluation of DA-9801, a new herbal drug for diabetic neuropathy, on metabolism-mediated interaction.
Archives of pharmacal research (2013-02-02)
Hye Young Ji, Kwang Hyeon Liu, Tae Yeon Kong, Hyeon-Uk Jeong, Sang-Zin Choi, Miwon Son, Yong-Yeon Cho, Hye Suk Lee
ABSTRACT
DA-9801, the mixture extract of Dioscoreae rhizoma and Dioscorea nipponica Makino, is a new herbal drug currently being evaluated in a phase II clinical study for the treatment of diabetic peripheral neuropathy in Korea. The inhibitory potentials of DA-9801, D. rhizoma extract, D. nipponica Makino extract, and dioscin, an active component of DA-9801, on eight human cytochrome P450 (CYP) enzymes and four UDP-glucuronosyltransferase (UGT) enzymes were investigated in human liver microsomes using liquid chromatography-tandem mass spectrometry. DA-9801 showed slight inhibition of CYP1A2, CYP2C8, UGT1A1, and UGT1A9 enzyme activities with IC(50) values of 396.4, 449.9, 226.0, and 408.8 μg/mL, respectively. D. rhizoma extract showed negligible inhibition of CYP and UGT activities, but D. nipponica extract slightly inhibited CYP1A2, CYP2C8, CYP2C9, UGT1A1, and UGT1A9 activities with IC(50) values of 264.2, 237.1, 206.8, 302.4, and 383.1 μg/mL, respectively. DA-9801 showed volume per dose index values of 0.44-0.88 L for a 200-mg dose, suggesting that they may not cause the inhibition of the metabolism of CYP1A2, CYP2C8, UGT1A1, and UGT1A9-catalyzed drugs in humans. These results suggest that the administration of DA-9801 in human may not cause clinically relevant inhibition of these enzymes.