MilliporeSigma
  • Tapasin-related protein TAPBPR is an additional component of the MHC class I presentation pathway.
Tapasin-related protein TAPBPR is an additional component of the MHC class I presentation pathway.
Proceedings of the National Academy of Sciences of the United States of America (2013-02-13)
Louise H Boyle, Clemens Hermann, Jessica M Boname, Keith M Porter, Peysh A Patel, Marian L Burr, Lidia M Duncan, Michael E Harbour, David A Rhodes, Karsten Skjødt, Paul J Lehner, John Trowsdale
ABSTRACT
Tapasin is an integral component of the peptide-loading complex (PLC) important for efficient peptide loading onto MHC class I molecules. We investigated the function of the tapasin-related protein, TAPBPR. Like tapasin, TAPBPR is widely expressed, IFN-γ-inducible, and binds to MHC class I coupled with β2-microglobulin in the endoplasmic reticulum. In contrast to tapasin, TAPBPR does not bind ERp57 or calreticulin and is not an integral component of the PLC. β2-microglobulin is essential for the association between TAPBPR and MHC class I. However, the association between TAPBPR and MHC class I occurs in the absence of a functional PLC, suggesting peptide is not required. Expression of TAPBPR decreases the rate of MHC class I maturation through the secretory pathway and prolongs the association of MHC class I on the PLC. The TAPBPR:MHC class I complex trafficks through the Golgi apparatus, demonstrating a function of TAPBPR beyond the endoplasmic reticulum/cis-Golgi. The identification of TAPBPR as an additional component of the MHC class I antigen-presentation pathway demonstrates that mechanisms controlling MHC class I expression remain incompletely understood.
Sigma-Aldrich
Protein Disulfide Isomerase from bovine liver, ≥100 units/mg protein, lyophilized powder