MilliporeSigma
  • Matrix metalloproteinases 2 and 9 as diagnostic markers in the progression to Chagas cardiomyopathy.
Matrix metalloproteinases 2 and 9 as diagnostic markers in the progression to Chagas cardiomyopathy.
American heart journal (2013-03-30)
Norma Leticia Bautista-López, Carlos A Morillo, Patricio López-Jaramillo, Roberto Quiroz, Carlos Luengas, Sandra Y Silva, Jacques Galipeau, Manoj Mathew Lalu, Richard Schulz
ABSTRACT
Infection with the Trypanosoma cruzi parasite is endemic in parts of Central and South America. Approximately 30% of those infected develop Chagas cardiomyopathy, the most common cause of heart failure in this region. No suitable biomarker is available that reflects the evolution of the disease. Although there is substantial evidence of a strong inflammatory reaction following infection that could activate matrix metalloproteinases (MMPs), their role in the development of Chagas cardiomyopathy is unknown. A cross-sectional study was conducted in Bucaramanga, Colombia, from 2002 to 2006, including 144 patients at different stages of Chagas disease and 44 control patients. The potential enzyme activities of MMP-2 and MMP-9 in plasma samples were determined by gelatin zymography. Clinical data including T cruzi serology, electrocardiograms, and echocardiograms were recorded for all patients. Densitometric analysis of potential enzyme activities in plasma samples showed a significant increase of 72-kd MMP-2 (P < .001) and 92-kd MMP-9 (P < .001) in T cruzi seropositive patients compared with control subjects. Matrix metalloproteinase 9 showed significantly increased activity in patients with abnormal electrocardiogram (P < .004) and with dilated cardiomyopathy compared (P < .001) with controls. Analysis of the MMP-2 and MMP-9 results in relation to clinical data revealed that abnormal heart relaxation correlated positively with high MMP-2 levels in patients with dilated cardiomyopathy (r = 0.75, P < .01). Plasma MMP-2 and MMP-9 both appear to be useful biomarkers for detecting the advent and progression of cardiomyopathy in T cruzi-infected individuals.