MilliporeSigma
  • Polymerase chain reaction coupling with magnetic nanoparticles-based biotin-avidin system for amplification of chemiluminescent detection signals of nucleic acid.
Polymerase chain reaction coupling with magnetic nanoparticles-based biotin-avidin system for amplification of chemiluminescent detection signals of nucleic acid.
Journal of nanoscience and nanotechnology (2011-04-02)
Zhiyang Li, Lei He, Nongyue He, Yan Deng, Zhiyang Shi, Hua Wang, Song Li, Hongna Liu, Zhifei Wang, Daxin Wang
ABSTRACT
A novel method was established through the detection of chemiluminescent signals of nucleic acid hybridization based on magnetic nanoparticles (MNPs) and PCR. 5' amino- modified specific probes were immobilized on the surface of silanized MNPs by Schiff reaction between amino and aldehyde group. The probes were used to capture the synthetic biotin-dUTP-labeled DNA fragments which were obtained by polymerase chain reaction (PCR). Then these complexes were bonded with streptavidin-modified alkaline phosphatase (SA-AP). Finally the chemiluminescent signals were detected by adding 3-(2'-spiroadamantane)- 4-methoxy -4-(3"-phosphoryloxy) phenyl-1, 2-dioxetane (AMPPD) which was the substrate reagent of AP. The concentration of probes which were immobilized on the surface of MNPs was studied, how to reduce the adsorption of SA-AP on the surface of MNPs was also researched. It was shown that 12.5 pmol of probes were immobilized on 1 mg of MNPs. Aldehyde-MNPs modified with probes could adsorb SA-AP, affecting the sensitivity of chemiluminescene consequently. Reduction of aldehyde group by sodium borohydride and blocking the bare position of MNPs with bovine serum albumin (BSA) could decrease the background of chemiluminescence, and this method has good specificity in detection of chloramphenicol acetyltransferase (CAT) gene.