MilliporeSigma
  • Synthesis and fucosidase inhibitory study of unnatural pyrrolidine alkaloid 4-epi-(+)-codonopsinine.
Synthesis and fucosidase inhibitory study of unnatural pyrrolidine alkaloid 4-epi-(+)-codonopsinine.
The Journal of organic chemistry (2011-04-19)
Alexis Kotland, Fabien Accadbled, Koen Robeyns, Jean-Bernard Behr
ABSTRACT
The total synthesis of enantiopure 4-epi-(+)-codonopsinine was achieved in 10 steps starting from D-ribose as a chiral building block. The key step involved a highly stereoselective nucleophilic addition of a Grignard reagent to a protected ribosylamine. Synthesis of the N-desmethyl derivative and its p-tolyl analogue was also accomplished, and the compounds were assayed against α-fucosidase.