MilliporeSigma
  • Randomised clinical trial: pre-dosing with taribavirin before starting pegylated interferon vs. standard combination regimen in treatment-naïve patients with chronic hepatitis C genotype 1.
Randomised clinical trial: pre-dosing with taribavirin before starting pegylated interferon vs. standard combination regimen in treatment-naïve patients with chronic hepatitis C genotype 1.
Alimentary pharmacology & therapeutics (2012-06-20)
M Palmer, R Rubin, V Rustgi
ABSTRACT
Combination therapy with the ribavirin (RBV) prodrug taribavirin (TBV) and pegylated interferon (PIFN) has produced lower rates of anaemia than with RBV and PIFN. Studies have demonstrated that the sharpest decline in viral load during TBV therapy occurs at Weeks 4 through 6, when TBV reaches steady-state blood levels. The current proof-of-concept study was conducted to examine whether first-order viral kinetics could be influenced by pre-dosing TBV to steady state before introducing PIFN. Therapy-naïve patients with chronic hepatitis C virus (HCV) genotype 1 (G1) were randomised to receive (i) TBV 600 mg BID monotherapy for 4 weeks followed by combination therapy with PIFN [pre-dosing arm (n = 23)] or (ii) TBV administered concurrently with PIFN [standard dosing arm (n = 19)]. More patients achieved undetectable virus or a ≥2-log(10) reduction of HCV RNA at Week 4 in the pre-dosing vs. the standard dosing arm [33% vs. 22% (P = 0.497)]. There was also a trend towards greater reduction in mean log(10) change in HCV RNA in the pre-dosing vs. the standard dosing arm, which was statistically significant at Day 1 [-0.34 ± 0.46 vs. 0.09 ± 0.32 (P < 0.003)] but not at other time points up to Week 24. No significant difference was observed in the rates of anaemia (haemoglobin <10 g/dL) between study arms (4.5% vs. 5.3%). Pre-dosing TBV prior to starting PIFN produces a trend towards improved efficacy although statistical significance was not reached in this small patient population. These results warrant larger clinical trials of TBV pre-dosing.
Sigma-Aldrich
Nucleoside Phosphorylase bacterial, recombinant, expressed in E. coli, ≥10 units/mg protein