MilliporeSigma
  • Involvement of the oxidative pentose phosphate pathway in thiamine biosynthesis in Salmonella typhimurium.
Involvement of the oxidative pentose phosphate pathway in thiamine biosynthesis in Salmonella typhimurium.
Journal of bacteriology (1996-03-01)
J L Enos-Berlage, D M Downs
ABSTRACT
purF mutants of Salmonella typhimurium are known to require a source of both purine and thiamine; however, exogenous pantothenate may be substituted for the thiamine requirement. We show here that the effect of pantothenate is prevented by blocks in the oxidative pentose phosphate pathway, gnd (encoding gluconate 6-phosphate [6-P] dehydrogenase) or zwf (encoding glucose 6-P dehydrogenase). We further show that the defects caused by these mutations can be overcome by increasing ribose 5-P, suggesting that ribose 5-P may play a role in the ability of pantothenate to substitute for thiamine.
Sigma-Aldrich
6-Phosphogluconic Dehydrogenase from yeast, lyophilized powder, ≥3.0 units/mg solid