MilliporeSigma
  • 1-Aminoindan-1,5-dicarboxylic acid: a novel antagonist at phospholipase C-linked metabotropic glutamate receptors.
1-Aminoindan-1,5-dicarboxylic acid: a novel antagonist at phospholipase C-linked metabotropic glutamate receptors.
Journal of medicinal chemistry (1995-09-15)
R Pellicciari, R Luneia, G Costantino, M Marinozzi, B Natalini, P Jakobsen, A Kanstrup, G Lombardi, F Moroni, C Thomsen
Sigma-Aldrich
AIDA, solid