MilliporeSigma
  • Structural characterization of nitric oxide synthase isoforms reveals striking active-site conservation.
Structural characterization of nitric oxide synthase isoforms reveals striking active-site conservation.
Nature structural biology (1999-03-13)
T O Fischmann, A Hruza, X D Niu, J D Fossetta, C A Lunn, E Dolphin, A J Prongay, P Reichert, D J Lundell, S K Narula, P C Weber
ABSTRACT
Crystal structures of human endothelial nitric oxide synthase (eNOS) and human inducible NOS (iNOS) catalytic domains were solved in complex with the arginine substrate and an inhibitor S-ethylisothiourea (SEITU), respectively. The small molecules bind in a narrow cleft within the larger active-site cavity containing heme and tetrahydrobiopterin. Both are hydrogen-bonded to a conserved glutamate (eNOS E361, iNOS E377). The active-site residues of iNOS and eNOS are nearly identical. Nevertheless, structural comparisons provide a basis for design of isozyme-selective inhibitors. The high-resolution, refined structures of eNOS (2.4 A resolution) and iNOS (2.25 A resolution) reveal an unexpected structural zinc situated at the intermolecular interface and coordinated by four cysteines, two from each monomer.
Sigma-Aldrich
Nitric Oxide Synthase, Inducible from mouse, recombinant, expressed in E. coli, buffered aqueous solution