MilliporeSigma
  • Read-through transcript from NM23-H1 into the neighboring NM23-H2 gene encodes a novel protein, NM23-LV.
Read-through transcript from NM23-H1 into the neighboring NM23-H2 gene encodes a novel protein, NM23-LV.
Genomics (2006-01-31)
Linda J Valentijn, Jan Koster, Rogier Versteeg
ABSTRACT
NM23-H1 and NM23-H2 are neighboring genes on chromosome 17q. They encode nucleoside diphosphate kinases that have additional roles in signal transduction, transcription, and apoptosis. NM23-H1 expression is a strong marker for prognosis and metastatic behavior in many tumor types. A new bioinformatic tool, TranscriptView, identified read-through transcripts that start in the NM23-H1 gene and continue in the neighboring NM23-H2 gene. Splicing results in a transcript containing exons 1 to 4 of NM23-H1 and exons 2 to 5 of NM23-H2. The resulting mRNA encodes a novel and long variant of the NM23 protein family, NM23-LV, which contains part of NM23-H1 and the complete NM23-H2 protein. The transcript was amplified and sequenced from two neuroblastoma cell lines, confirming the presence of the predicted NM23-LV mRNA in vivo. Tissue analysis showed that NM23-LV is ubiquitously expressed, with the exception of the kidney. Neuroblastoma tumors show high-level expression of NM23-H1 and-H2 as well as NM23-LV mRNA. In neuroblastoma cells, the NM23-LV protein has mainly a cytoplasmic localization, but some nuclear staining was observed as well.
Sigma-Aldrich
Nucleoside 5′-Diphosphate Kinase from bovine liver, buffered aqueous glycerol solution, ≥1,000 units/mg protein (biuret)