MilliporeSigma
  • In vitro activity and preclinical profile of TMC435350, a potent hepatitis C virus protease inhibitor.
In vitro activity and preclinical profile of TMC435350, a potent hepatitis C virus protease inhibitor.
Antimicrobial agents and chemotherapy (2009-01-28)
Tse-I Lin, Oliver Lenz, Gregory Fanning, Thierry Verbinnen, Frédéric Delouvroy, Annick Scholliers, Katrien Vermeiren, Asa Rosenquist, Michael Edlund, Bertil Samuelsson, Lotta Vrang, Herman de Kock, Piet Wigerinck, Pierre Raboisson, Kenneth Simmen
ABSTRACT
The hepatitis C virus (HCV) NS3/4A serine protease has been explored as a target for the inhibition of viral replication in preclinical models and in HCV-infected patients. TMC435350 is a highly specific and potent inhibitor of NS3/4A protease selected from a series of novel macrocyclic inhibitors. In biochemical assays using NS3/4A proteases of genotypes 1a and 1b, inhibition constants of 0.5 and 0.4 nM, respectively, were determined. TMC435350 inhibited HCV replication in a cellular assay (subgenomic 1b replicon) with a half-maximal effective concentration (EC(50)) of 8 nM and a selectivity index of 5,875. The compound was synergistic with alpha interferon and an NS5B inhibitor in the replicon model and additive with ribavirin. In rats, TMC435350 was extensively distributed to the liver and intestinal tract (tissue/plasma area under the concentration-time curve ratios of >35), and the absolute bioavailability was 44% after a single oral administration. Compound concentrations detected in both plasma and liver at 8 h postdosing were above the EC(99) value measured in the replicon. In conclusion, given the selective and potent in vitro anti-HCV activity, the potential for combination with other anti-HCV agents, and the favorable pharmacokinetic profile, TMC435350 has been selected for clinical development.
Sigma-Aldrich
Ribavirin, antiviral
Simeprevir, ≥98% (HPLC)