MilliporeSigma
  • Derivatives of 1-(1,3-benzodioxol-5-yl)-2-butanamine: representatives of a novel therapeutic class.
Derivatives of 1-(1,3-benzodioxol-5-yl)-2-butanamine: representatives of a novel therapeutic class.
Journal of medicinal chemistry (1986-10-01)
D E Nichols, A J Hoffman, R A Oberlender, P Jacob, A T Shulgin
ABSTRACT
The alpha-ethyl phenethylamine derivative 1-(1,3-benzodioxol-5-yl)-2-butanamine was prepared. An asymmetric synthesis was used to prepare the enantiomers of this compound and the related alpha-methyl homologue (MDA). The racemates and enantiomers of both compounds were evaluated in the two-lever drug discrimination assay in rats trained to discriminate saline from 0.08 mg/kg of LSD tartrate. Stimulus generalization occurred with the racemate and the R-(-) enantiomer of the alpha-methyl homologue and the S-(+) enantiomer of the alpha-ethyl primary amine. No generalization occurred with the other enantiomers or with the N-methyl derivatives of either series. Human psychopharmacology studies revealed that the N-methyl derivative of the title compound was nonhallucinogenic and that it had a new, novel psychoactive effect. It is suggested that this compound is the prototype of a new pharmacologic class that may have value in facilitating psychotherapy and that this class be designated as entactogens.