MilliporeSigma
  • The human sorbitol dehydrogenase gene: cDNA cloning, sequence determination, and mapping by fluorescence in situ hybridization.
The human sorbitol dehydrogenase gene: cDNA cloning, sequence determination, and mapping by fluorescence in situ hybridization.
Genomics (1994-05-15)
F K Lee, M C Cheung, S Chung
ABSTRACT
The cDNA for human sorbitol dehydrogenase (SORD) has been cloned and sequenced. It translates into a peptide of 356 amino acid residues, one more than the sequence previously reported from peptide analysis. An extra alanine was found at the acetyl-blocked N-terminal, between positions 1 and 4. This matches the rat cDNA, which also has 356 amino acids, with an extra proline at position 3. Four other mis-matches were also observed, but these are all amino acid substitutions that occur outside proposed functionally important regions. Further work must be performed to determine whether these discrepancies represent polymorphic forms of the enzyme. The SORD gene was mapped by fluorescence in situ hybridization and found to occupy a single site on chromosome 15q15, indicating that it is a single-copy gene. This was confirmed by Southern blot hybridization. SORD is thought to be involved in the etiology of diabetic complications, and its deficiency has been linked to congenital cataracts. The cloned gene could be used as a probe to study the role of this enzyme in the pathogenesis of these diseases.