MilliporeSigma
  • Involvement of the clock gene Rev-erb alpha in the regulation of glucagon secretion in pancreatic alpha-cells.
Involvement of the clock gene Rev-erb alpha in the regulation of glucagon secretion in pancreatic alpha-cells.
PloS one (2013-08-13)
Elaine Vieira, Laura Marroquí, Ana Lucia C Figueroa, Beatriz Merino, Rebeca Fernandez-Ruiz, Angel Nadal, Thomas P Burris, Ramon Gomis, Ivan Quesada
ABSTRACT
Disruption of pancreatic clock genes impairs pancreatic beta-cell function, leading to the onset of diabetes. Despite the importance of pancreatic alpha-cells in the regulation of glucose homeostasis and in diabetes pathophysiology, nothing is known about the role of clock genes in these cells. Here, we identify the clock gene Rev-erb alpha as a new intracellular regulator of glucagon secretion. Rev-erb alpha down-regulation by siRNA (60-70% inhibition) in alphaTC1-9 cells inhibited low-glucose induced glucagon secretion (p<0.05) and led to a decrease in key genes of the exocytotic machinery. The Rev-erb alpha agonist GSK4112 increased glucagon secretion (1.6 fold) and intracellular calcium signals in alphaTC1-9 cells and mouse primary alpha-cells, whereas the Rev-erb alpha antagonist SR8278 produced the opposite effect. At 0.5 mM glucose, alphaTC1-9 cells exhibited intrinsic circadian Rev-erb alpha expression oscillations that were inhibited by 11 mM glucose. In mouse primary alpha-cells, glucose induced similar effects (p<0.001). High glucose inhibited key genes controlled by AMPK such as Nampt, Sirt1 and PGC-1 alpha in alphaTC1-9 cells (p<0.05). AMPK activation by metformin completely reversed the inhibitory effect of glucose on Nampt-Sirt1-PGC-1 alpha and Rev-erb alpha. Nampt inhibition decreased Sirt1, PGC-1 alpha and Rev-erb alpha mRNA expression (p<0.01) and glucagon release (p<0.05). These findings identify Rev-erb alpha as a new intracellular regulator of glucagon secretion via AMPK/Nampt/Sirt1 pathway.
Sigma-Aldrich
GSK4112, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
SR8278, ≥98% (HPLC)
Pricing and availability is not currently available.