MilliporeSigma
c-Met inhibitors.
Infectious agents and cancer (2013-04-10)
Anum Mughal, Hafiz Muhammad Aslam, Asfandyar Sheikh, Agha Muhammad Hammad Khan, Shafaq Saleem
ABSTRACT
c-Met is a receptor tyrosine kinase that encodes protein such as hepatocyte growth factor receptor (HGFR). Inappropriate activity of c-Met can cause wide variety of carcinomas. c-Met inhibitor are relatively new class of small molecules that inhibit the enzymatic activity of c-Met tyrosine kinase. Met inhibitors divided into two main classes: class I (SU-11274-like) and class II (AM7-like). The use of c-Met inhibitors with other therapeutic agents could be crucial for overcoming potential resistance as well as for improving overall clinical benefit. Met pathway inhibitors might be used in combination with other treatments, including chemo-, radio- or immunotherapy.