MilliporeSigma
CASH, a novel caspase homologue with death effector domains.
The Journal of biological chemistry (1997-08-08)
Y V Goltsev, A V Kovalenko, E Arnold, E E Varfolomeev, V M Brodianskii, D Wallach
ABSTRACT
CASP-8 and CASP-10, members of a cysteine protease family that participates in apoptosis, interact with MORT1/FADD, an adapter protein in the CD120a (p55 tumor necrosis factor receptor), and CD95 (Fas/Apo-1) death-inducing signaling pathways, through a shared N-terminal sequence motif, the death effector domain. We report cloning of two splice variants of a novel protein, CASH, that contain two N-terminal death effector domains and can bind through them to each other, to MORT1/FADD, to CASP-8, and to CASP-10. The unique C-terminal part of the longer variant shows marked sequence homology to the caspase protease region yet lacks several of the conserved caspase active site residues, suggesting that it is devoid of cysteine protease activity. Overexpression of the short CASH splice variant strongly inhibited cytotoxicity induction by CD120a and CD95. Expression of the longer variant, while inhibiting cytotoxicity in HeLa cells, had a marked cytocidal effect in 293 cells that could be shown to involve its protease homology region. The findings suggest that CASH acts as an attenuator and/or initiator in CD95 and CD120a signaling for cell death.