MilliporeSigma
  • Evidence for the AT1 subtype of the angiotensin II receptor in the rat submandibular gland.
Evidence for the AT1 subtype of the angiotensin II receptor in the rat submandibular gland.
Biological & pharmaceutical bulletin (2000-10-21)
S Matsubara, K Saito, Y Kizawa, M Sano, M Osawa, K Iwamoto, H Murakami
ABSTRACT
We have characterized angiotensin II (Ang II) receptor subtypes on rat submandibular gland membranes using a radioligand binding assay. [3H]Ang II binding to the membrane fractions exhibited both high (Kd =0.08 nm, Bmax =2.19 fmol/mg protein) and low (Kd =4.19 nm, Bmax = 13.7 fmol/mg protein) affinity. Ang 11, Ang III and saralasin completely displaced the [3H]Ang II binding, whereas CV-11974, an AT1 receptor antagonist and PD123319, an AT2 receptor antagonist maximally displaced up to approximately 87 and 13% of the total binding, respectively. [3H]DuP753 binding to the membrane fractions exhibited a single population of binding site with a Kd of 4.22 nM and Bmax of 3.77 pmol/mg protein. Ang II, Ang III and CV-11974 completely displaced the [3H]DuP753 binding with slope factors near unity, but PD123319 did not. These findings suggest that rat submandibular gland membranes contain predominantly the AT1 receptor subtype.