MilliporeSigma
  • The murine ufo receptor: molecular cloning, chromosomal localization and in situ expression analysis.
The murine ufo receptor: molecular cloning, chromosomal localization and in situ expression analysis.
Oncogene (1992-07-01)
M Faust, C Ebensperger, A S Schulz, L Schleithoff, H Hameister, C R Bartram, J W Janssen
ABSTRACT
We have cloned the mouse homologue of the ufo oncogene. It encodes a novel tyrosine kinase receptor characterized by a unique extracellular domain containing two immunoglobulin-like and two fibronectin type III repeats. Comparison of the predicted ufo amino acid sequences of mouse and man revealed an overall identity of 87.6%. The ufo locus maps to mouse chromosome 7A3-B1 and thereby extends the known conserved linkage group between mouse chromosome 7 and human chromosome 19. RNA in situ hybridization analysis established the onset of specific ufo expression in the late embryogenesis at day 12.5 post coitum (p.c.) and localized ufo transcription to distinct substructures of a broad spectrum of developing tissues (e.g. subepidermal cells of the skin, mesenchymal cells of the periosteum). In adult animals ufo is expressed in cells forming organ capsules as well as in connective tissue structures. ufo may function as a signal transducer between specific cell types of mesodermal origin.