MilliporeSigma
  • A high throughput assay for inhibitors of HIV-1 protease. Screening of microbial metabolites.
A high throughput assay for inhibitors of HIV-1 protease. Screening of microbial metabolites.
FEBS letters (1991-02-25)
E Sarubbi, M L Nolli, F Andronico, S Stella, G Saddler, E Selva, A Siccardi, M Denaro
ABSTRACT
A novel method for discovery of HIV-1 protease inhibitors in complex biological samples has been developed. The assay is based on two specific reagents: a recombinant protein constituted by a portion of the HIV-1 Gag polyprotein comprising the p17-p24 cleavage site, fused to E. coli beta-galactosidase, and a monoclonal antibody which binds the fusion protein in the Gag region. Binding occurs only if the fusion protein has not been cleaved by the HIV-1 protease. The assay has been adapted for the screening of large numbers of samples in standard 96-well microtiter plates. Using this method about 12000 microbial fermentation broths have been tested and several HIV-1 protease inhibitory activities have been detected. One of these has been studied in detail.