MilliporeSigma
  • New insights into the control of cell growth; the role of the AxI family.
New insights into the control of cell growth; the role of the AxI family.
Pathology (1997-05-01)
K E Crosier, P S Crosier
ABSTRACT
AxI, Dtk and Mer are recently described receptors that constitute a new receptor tyrosine kinase subfamily. They bind the vitamin K-dependent protein growth-arrest-specific gene 6 (Gas6) that is structurally related to the anticoagulation factor protein S. Studies suggest a role for these receptors in developmental processes. In the function of the hematopoietic and nervous systems and in tumorigenesis.