MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Matrix solid-phase dispersion microextraction and determination by high-performance liquid chromatography with UV detection of pesticide residues in citrus fruit.
Matrix solid-phase dispersion microextraction and determination by high-performance liquid chromatography with UV detection of pesticide residues in citrus fruit.
Journal of chromatography. A (1999-05-18)
A I Valenzuela, R Lorenzini, M J Redondo, G Font
ABSTRACT
A multiresidue method based on matrix solid-phase dispersion (MSPD) microextraction was studied to determine the carbamate, benfuracarb, and urea insecticides, diflubenzuron, flufenoxuron hexaflumuron and hexythiazox, used in control of citrus pests. Optimisation of different parameters, such as the type of solid support for matrix dispersion, elution solvents and the clean-up step were carried out. The method used 0.5 g of orange sample, C8 bonded silica as MSPD sorbent and dichloromethane as eluting solvent. Recoveries, at spiked concentrations below the maximum residue levels established by Spanish Government, were between 74 and 84% with relative standard deviations ranging from 2 to 4%. The limits of quantification were from 0.15 to 0.25 microgram/g using high-performance liquid chromatography with UV detection at 200 nm. The method may be useful as a screening protocol for the determination of these newly developed pesticides in citrus samples.
Product Description
Supelco
Benfuracarb, PESTANAL®, analytical standard
Supelco
Flufenoxuron, PESTANAL®, analytical standard