MilliporeSigma
  • New insights into the pyrimidine salvage pathway of Saccharomyces cerevisiae: requirement of six genes for cytidine metabolism.
New insights into the pyrimidine salvage pathway of Saccharomyces cerevisiae: requirement of six genes for cytidine metabolism.
Current genetics (1999-09-29)
J E Kurtz, F Exinger, P Erbs, R Jund
ABSTRACT
Cytidine metabolism in the yeast Saccharomyces cerevisiae was analyzed by genetic and biochemical approaches. Disruption of a unique ORF (Genbank accession No. U 20865) bearing homology with eucaryotic or bacterial cytidine deaminases abolished cytidine deaminase activity and resulted in 5-fluorocytidine resistance. The gene encoding cytidine deaminase will be referred to as CDD1 (Genbank accession number AF080089). The ability to isolate mutants resistant to 5-fluorocytidine which mapped to five other loci demonstrated the existence of a complex cytidine metabolic network. Deciphering this network revealed several original features:(1) cytidine entry is mediated by the purine-cytosine transporter (Fcy2p),(2) cytidine is cleaved into cytosine by the uridine nucleosidase (Urh1p),(3) cytidine is phosphorylated into CMP by the uridine kinase (Urk1p),(4) a block in cytosine deaminase (Fcy1p), but not in cytidine deaminase (Cdd1p), constitutes a limiting step in cytidine utilisation as a UMP precursor.