MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Distribution of bromophenols in species of marine polychaetes and bryozoans from eastern Australia and the role of such animals in the flavor of edible ocean fish and prawns (shrimp).
Distribution of bromophenols in species of marine polychaetes and bryozoans from eastern Australia and the role of such animals in the flavor of edible ocean fish and prawns (shrimp).
Journal of agricultural and food chemistry (1999-12-20)
F B Whitfield, M Drew, F Helidoniotis, D Svoronos
ABSTRACT
Sixteen species (44 samples) of marine polychaetes and 10 species (14 samples) of bryozoans from eastern Australia were analyzed by GC/MS for the key seafood flavor components 2- and 4-bromophenol, 2, 4- and 2,6-dibromophenol, and 2,4,6-tribromophenol. All five bromophenols were found in 91% of polychaetes and 64% of bryozoans. The remaining samples all contained at least three bromophenols. 2,4, 6-Tribromophenol was found in all polychaetes and bryozoans and, with few exceptions, was present in the highest concentrations. The total bromophenol content determined on a wet-weight basis varied widely between species: for polychaetes, from 58 ng/g for Australonuphis teres to 8.3 million ng/g for Barantolla lepte, and for bryozoans, from 36 ng/g for Cladostephus spongiosus to 1668 ng/g for Amathia wilsoni. Species of polychaetes with the highest concentrations of bromophenols were all collected from muddy environments. The possible effects that dietary polychaetes and bryozoans have on the ocean-, brine-, or iodine-like flavors of certain seafoods are discussed.
Sigma-Aldrich
2,6-Dibromophenol, 99%
Sigma-Aldrich
2,4-Dibromophenol, 95%
Sigma-Aldrich
2-Bromophenol, 98%
Sigma-Aldrich
4-Bromophenol, 99%