MilliporeSigma
  • The anticonvulsive effect of non-NMDA antagonist GYKI 52466 on 3-aminopyridine-induced primary and secondary cortical ictal activity in rat.
The anticonvulsive effect of non-NMDA antagonist GYKI 52466 on 3-aminopyridine-induced primary and secondary cortical ictal activity in rat.
Acta biologica Hungarica (1999-11-26)
B Barna, M Szente, A Szász
ABSTRACT
The effect of GYKI 52466, a selective, non-competitive antagonist of the AMPA glutamate receptor subtype was investigated on the development, expression and propagation of 3-aminopyridine-induced cortical ictal activity, both in the primary and secondary focus. In one group of animals GYKI 52466 was administered intraperitoneally, 20 minutes prior to the local application of the convulsant the surface of the cortex of anaesthetized rats. Control animals were injected by physiological solution. Different parameters of electrographic ictal discharges were measured under the influence of the antagonist and compared to control values. The results demonstrate that GYKI 52466 exerts anticonvulsive effects on both the induction and the expression of primary and secondary electrical ictal activity, by delaying the onset of the first ictal period, shortening the duration of ictal activity and decreasing the amplitudes of epileptiform discharges. However, the seizure propagation was not influenced significantly. It is suggested that the initiation, maintenance and the propagation of spontaneous seizures may be controlled by separate mechanisms and that changes can occur in one of the procedures without parallel changes in others. The observations of the present study extend those reported previously by others, namely that activation of non-NMDA receptors is significantly involved in the initiation and maintenance of cortical epileptiform activity.
Sigma-Aldrich
3-Aminopyridine, 99%