MilliporeSigma
  • Dietary flavonoid and isoflavone glycosides are hydrolysed by the lactase site of lactase phlorizin hydrolase.
Dietary flavonoid and isoflavone glycosides are hydrolysed by the lactase site of lactase phlorizin hydrolase.
FEBS letters (2000-02-29)
A J Day, F J Cañada, J C Díaz, P A Kroon, R Mclauchlan, C B Faulds, G W Plumb, M R Morgan, G Williamson
ABSTRACT
Lactase phlorizin hydrolase (LPH; EC 3.2.1.62) is a membrane-bound, family 1 beta-glycosidase found on the brush border of the mammalian small intestine. LPH, purified from sheep small intestine, was capable of hydrolysing a range of flavonol and isoflavone glycosides. The catalytic efficiency (k(cat)/K(m)) for the hydrolysis of quercetin-4'-glucoside, quercetin-3-glucoside, genistein-7-glucoside and daidzein-7-glucoside was 170, 137, 77 and 14 (mM(-1) s(-1)) respectively. The majority of the activity occurred at the lactase and not phlorizin hydrolase site. The ability of LPH to deglycosylate dietary (iso)flavonoid glycosides suggests a possible role for this enzyme in the metabolism of these biologically active compounds.