MilliporeSigma
Piezoreflectance spectra of 1s Z12 exciton in CuBr.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2000-03-23)
H Erguig, O Pages, A Zaoui, J P Laurenti, M Certier, B Kabouchi
ABSTRACT
We have investigated the piezoreflectance spectra (deltaR/R) of the 1s Z12 exciton in single crystals of copper bromide CuBr at the temperature of 95 K with linearly polarized light. The spectra were successively studied with the pressure p along the [001] and [111] axis and the wave vector k of the incident light parallel to the [110] direction. The shear deformation potentials b and d of the 1s Z12 exciton were deduced from the stress-induced shift and splitting. The results are compared with those obtained in other works.
Sigma-Aldrich
Copper(II) bromide, 99%
Sigma-Aldrich
Copper(II) bromide, 99.999% trace metals basis