MilliporeSigma
  • Liver subcellular fractions from rats treated by organosulfur compounds from Allium modulate mutagen activation.
Liver subcellular fractions from rats treated by organosulfur compounds from Allium modulate mutagen activation.
Mutation research (2001-02-07)
D Guyonnet, C Belloir, M Suschetet, M H Siess, A M Le Bon
ABSTRACT
The effects of in vivo administration of naturally occurring organosulfur compounds (OSCs) from Allium species were studied on the activation of several mutagens. Male SPF Wistar rats were given p.o. one of either diallyl sulfide (DAS), diallyl disulfide (DADS), dipropyl sulfide (DPS) or dipropyl disulfide (DPDS) during 4 consecutive days and the ability of hepatic S9 and microsomes from treated rats to activate benzo[a]pyrene (BaP), cyclophosphamide (CP), dimethylnitrosamine (DMN), N-nitrosopiperidine (N-PiP) and 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) was determined in the Ames test. Administration of DAS, DPS and DPDS resulted in a significant increase of the activation of BaP, CP, N-PiP and PhIP mediated by S9 and microsomes while DADS treatment only increased the mutagenicity of PhIP. In contrast, S9 from DADS-treated rats significantly inhibited the mutagenicity of N-PiP and BaP. DAS, DADS and DPS strongly inhibited DMN mutagenicity while DPDS enhanced it. To understand the mechanisms underlying these effects, the modifications of the activities of specific isozymes of CYP involved in the activation of these mutagens were studied. DAS, DPS and DPDS strongly enhanced pentoxyresorufin O-dealkylase (PROD) activity related to CYP2B and slightly increased ethoxyresorufin O-deethylase (EROD) and methoxyresorufin O-demethylase (MROD) activities related to CYP1A family. DADS exerted the same effects than other OSCs but to a lesser extent. p-Nitrophenol hydroxylase (PNPH) activity related to CYP2E1 was inhibited by DAS and DADS, whereas DPDS significantly increased this activity. Hence, the effects of OSCs on the mutagenicity of several genotoxic compounds are mediated by modification (enhancement or inhibition) of specific CYP involved in their activation.
Sigma-Aldrich
Dipropyl sulfide, 97%
Supelco
1-Nitrosopiperidine solution, certified reference material, 5000 μg/mL in methanol