MilliporeSigma
  • Insight into acid-mediated asymmetric spirocyclization in the presence of a chiral diol.
Insight into acid-mediated asymmetric spirocyclization in the presence of a chiral diol.
Chemical & pharmaceutical bulletin (2000-10-25)
T Kiguchi, Y Tsurusaki, S Yamada, M Aso, M Tanaka, K Sakai, H Suemune
ABSTRACT
Asymmetric spirocyclization based on intramolecular conjugate addition using a combination of a Lewis acid and an optically active cyclohexane-1,2-diol has been studied in connection with 1) the effect of substituents on the cyclohexane-1,2-diol and 2) the effect of substituents on the substrate. This reaction was found to be both thermodynamically and kinetically controlled under restricted conditions.
Sigma-Aldrich
cis-1,2-Cyclohexanediol, 99%
Sigma-Aldrich
trans-1,2-Cyclohexanediol, 98%