MilliporeSigma
A chlorinated amide and piperidine alkaloids from Aloe sabaea.
Phytochemistry (2001-01-05)
T Blitzke, A Porzel, M Masaoud, J Schmidt
ABSTRACT
Phytochemical investigations of Aloe sabaea afforded a new chlorinated amide, N-4'-chlorobutylbutyramide, and the toxic piperidine alkaloids coniine, gamma-coniceine and the quarternary N,N-dimethylconiine. This is the first report of the occurrence of a chlorinated compound in the Aloeaceae family.