MilliporeSigma
  • N-acetylation and N-formylation of m-aminobenzoic acid by cell suspension cultures of Solanum laciniatum.
N-acetylation and N-formylation of m-aminobenzoic acid by cell suspension cultures of Solanum laciniatum.
Journal of Asian natural products research (2001-03-16)
A Syahrani, T S Panjaitan, G Indrayanto, A L Wilkins
ABSTRACT
Two new biotransformation products, N-acetyl-m-aminobenzoic acid and N-formyl-m-aminobenzoic acid were isolated from cell suspension cultures of Solanum laciniatum following administration of m-aminobenzoic acid, and their structures were elucidated using one- and two-dimensional 1H- and 13C-NMR data.
Sigma-Aldrich
3-Aminobenzoic acid, 98%