MilliporeSigma
Chemical constituents of Pseudopterogorgia species of the Indian ocean.
Journal of Asian natural products research (2001-05-18)
M Vanisree, G V Subbaraju, C B Rao
ABSTRACT
A new ceramide, (2S,3R,4E)-1,3-dihydroxy-2-[(hexadecanoyl)amino]-nonadeca-4-ene (1), cholest-5-en-3beta,7beta,19-triol (2), identified as its, peracetyl derivative (3), and batyl alcohol (4) were isolated from Pseudopterogorgia species. 1 exhibited antibacterial activity against Gram-positive and Gram-negative bacteria.
Sigma-Aldrich
Batyl alcohol, 99%