MilliporeSigma
Regioselective aromatic borylation in an inert solvent.
Organic letters (2001-09-01)
M K Tse, J Y Cho, M R Smith
ABSTRACT
[reaction: see text]. A protocol for performing Rh catalyzed aromatic borylations in cyclohexane has been devised. Borylation at the 5-position of several 1,3-substituted aromatic species ranging from electron-rich (1,3-(NMe(2))(2)C(6)H(4)) to electron-deficient (1,3-(CF(3))(2)C(6)H(4)) yields the corresponding aryl boronate esters. Veratrole was selectively borylated at the 4-position, thus extending regioselectivity to 1,2-substituted benzenes. Selective borylation at the 3-position of an N-protected pyrrole has also been demonstrated, providing a valuable reagent for cross-coupling reactions in a single step.
Sigma-Aldrich
1,2-Dimethoxybenzene, ≥99%, FG