MilliporeSigma
  • Effects of pyridaphenthion on growth of five freshwater species of phytoplankton. A laboratory study.
Effects of pyridaphenthion on growth of five freshwater species of phytoplankton. A laboratory study.
Chemosphere (2001-09-06)
C Sabater, J M Carrasco
ABSTRACT
The acute toxicity of the insecticide and acaricide pyridaphenthion to five species of freshwater phytoplankton, Scenedesmus acutus, Scenedesmus subspicatus, Chlorella vulgaris, Chlorella saccharophila and Pseudanabaena galeata was determined. Insecticide concentrations eliciting a 50% growth reduction over 96 h (EC50) ranged from 2.2 to 30.9 mg/l. The two species of Chlorella and the cyanobacteria P. galeata were more tolerant than the two species of Scenedesmus. Concentrations of pyridaphenthion detected in some natural waters were less than the toxic threshold for these species.