MilliporeSigma
  • Synthesis, topoisomerase I inhibition and antitumor cytotoxicity of 2,2':6',2"-, 2,2':6',3"- and 2,2':6',4"-terpyridine derivatives.
Synthesis, topoisomerase I inhibition and antitumor cytotoxicity of 2,2':6',2"-, 2,2':6',3"- and 2,2':6',4"-terpyridine derivatives.
Bioorganic & medicinal chemistry letters (2001-09-12)
L X Zhao, T S Kim, S H Ahn, T H Kim, E K Kim, W J Cho, H Choi, C S Lee, J A Kim, T C Jeong, C J Chang, E S Lee
ABSTRACT
For the development of new anticancer agents, 2,2':6',2"-, 2,2':6',3"- and 2,2':6',4"-terpyridine derivatives were designed and evaluated for their topoisomerase I inhibitory activity and antitumor cytotoxicity. Structure-activity relationship studies indicated that 2,2':6',2"-terpyridine derivatives were highly cytotoxic toward several human tumor cell lines, whereas 2,2':6',3"- and 2,2':6',4"-terpyridine derivatives were potent topoisomerase I inhibitors.
Sigma-Aldrich
2,2′:6′,2′′-Terpyridine, 98%