MilliporeSigma
  • Activity of cytisine and its brominated isosteres on recombinant human alpha7, alpha4beta2 and alpha4beta4 nicotinic acetylcholine receptors.
Activity of cytisine and its brominated isosteres on recombinant human alpha7, alpha4beta2 and alpha4beta4 nicotinic acetylcholine receptors.
Journal of neurochemistry (2001-09-13)
L M Houlihan, Y Slater, D L Guerra, J H Peng, Y P Kuo, R J Lukas, B K Cassels, I Bermudez
ABSTRACT
Effects of cytisine (cy), 3-bromocytisine (3-Br-cy), 5-bromocytisine (5-Br-cy) and 3,5-dibromocytisine (3,5-diBr-cy) on human (h) alpha7-, alpha4beta2- and alpha4beta4 nicotinic acetylcholine (nACh) receptors, expressed in Xenopus oocytes and cell lines, have been investigated. Cy and its bromo-isosteres fully inhibited binding of both [alpha-(125)I]bungarotoxin ([alpha-(125)I]BgTx) to halpha7- and [(3)H]cy to halpha4beta2- or halpha4beta4-nACh receptors. 3-Br-cy was the most potent inhibitor of both [alpha-(125)I]BgTx and [(3)H]cy binding. Cy was less potent than 3-Br-cy, but 5-Br-cy and 3,5-diBr-cy were the least potent inhibitors. Cy and 3-Br-cy were potent full agonists at halpha7-nACh receptors but behaved as partial agonists at halpha4beta2- and halpha4beta4-nACh receptors. 5-Br-cy and 3,5-diBr-cy had low potency and were partial agonists at halpha7- and halpha4beta4-nACh receptors, but they elicited no responses on halpha4beta2-nACh receptors. Cy and 3-Br-cy produced dual dose-response curves (DRC) at both halpha4beta2- and halpha4beta4-nACh receptors, but ACh produced dual DRC only at halpha4beta2-nACh receptors. Low concentrations of cy, 3-Br-cy and 5-Br-cy enhanced ACh responses of oocytes expressing halpha4beta2-nACh receptors, but at high concentrations they inhibited the responses. In contrast, 3,5-diBr-cy only inhibited, in a competitive manner, ACh responses of halpha4beta2-nACh receptors. It is concluded that bromination of the pyridone ring of cy produces marked changes in effects of cy that are manifest as nACh receptor subtype-specific differences in binding affinities and in functional potencies and efficacies.