MilliporeSigma
[Anti-corrosive effect of pesticides in soil corrosion conditions].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2001-11-06)
N V Smykun, A P Tretiak, I N Kurmakova
ABSTRACT
Effect of some sub-standard pesticides (Ramrod, Linuron, Simazin) with respect to corrosion-active groups of microorganisms: sulphate-reducing bacteria (SRB), denitrifying bacteria (DNB), saprophytic bacteria (SB) and their inhibiting properties under the conditions of active corrosion have been studied to estimate a possibility to use them as biocide additions when producing protective materials. It has been shown that the sub-standard pesticides Ramrod and Simazin are promising for to be used as the biocides additions under the protection of bioresistant materials. It is supposed that inhibitors-biocides may be found in a series of compounds obtained under chemical modification of substandard pesticides Ramrod and Simazin.
Product Description
Supelco
Propachlor, PESTANAL®, analytical standard