MilliporeSigma
  • [Isolation and identification of chemical constituents from Acropora pulchra].
[Isolation and identification of chemical constituents from Acropora pulchra].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2002-01-22)
Y Liu, S Xu, S Guo, X Li
ABSTRACT
Three compounds, N-1-(hydroxymethyl)-2-hydroxyl-(E,E)-3,7-heptadecadienal-hexadecanamide(1), batyl alcohol(2) and n-cetanol(3) were isolated from the EtOAc portion of the ethanol extract of the hard coral Acropora pulchra collected from north China sea, and their structures were determined by MS, 1HNMR, 13CNMR analysis.
Sigma-Aldrich
Batyl alcohol, 99%