MilliporeSigma
  • Three ginkgolide hydrates from Ginkgo biloba L.: ginkgolide A monohydrate, ginkgolide C sesquihydrate and ginkgolide J dihydrate, all determined at 120 K.
Three ginkgolide hydrates from Ginkgo biloba L.: ginkgolide A monohydrate, ginkgolide C sesquihydrate and ginkgolide J dihydrate, all determined at 120 K.
Acta crystallographica. Section C, Crystal structure communications (2002-03-01)
Jianping Zhao, Ilias Muhammad, D Chuck Dunbar, Ikhlas A Khan, Nikolaus H Fischer, Frank R Fronczek
ABSTRACT
A low-temperature structure of ginkgolide A monohydrate, (1R,3S,3aS,4R,6aR,7aR,7bR,8S,10aS,11aS)-3-(1,1-dimethylethyl)-hexahydro-4,7b-dihydroxy-8-methyl-9H-1,7a-epoxymethano-1H,6aH-cyclopenta[c]furo[2,3-b]furo[3',2':3,4]cyclopenta[1,2-d]furan-5,9,12(4H)-trione monohydrate, C(20)H(24)O(9) x H(2)O, obtained from Mo K alpha data, is a factor of three more precise than the previous room-temperature determination. A refinement of the ginkgolide A monohydrate structure with Cu K alpha data has allowed the assignment of the absolute configuration of the series of compounds. Ginkgolide C sesquihydrate, (1S,2R,3S,3aS,4R,6aR,7aR,7bR,8S,10aS,11S,11aR)-3-(1,1-dimethylethyl)-hexahydro-2,4,7b,11-tetrahydroxy-8-methyl-9H-1,7a-epoxymethano-1H,6aH-cyclopenta[c]furo[2,3-b]furo[3',2':3,4]cyclopenta[1,2-d]furan-5,9,12(4H)-trione sesquihydrate, C(20)H(24)O(11) x 1.5H(2)O, has two independent diterpene molecules, both of which exhibit intramolecular hydrogen bonding between OH groups. Ginkgolide J dihydrate, (1S,2R,3S,3aS,4R,6aR,7aR,7bR,8S,10aS,11aS)-3-(1,1-dimethylethyl)-hexahydro-2,4,7b-trihydroxy-8-methyl-9H-1,7a-epoxymethano-1H,6aH-cyclopenta[c]furo[2,3-b]furo[3',2':3,4]cyclopenta[1,2-d]furan-5,9,12(4H)-trione dihydrate, C(20)H(24)O(10) x 2H(2)O, has the same basic skeleton as the other ginkgolides, with its three OH groups having the same configurations as those in ginkgolide C. The conformations of the six five-membered rings are quite similar across ginkgolides A-C and J, except for the A and F rings of ginkgolide A.