MilliporeSigma
  • Identification of fonofos metabolites in Latuca sativa, Beta vulgaris, and Triticum aestivum by packed capillary flow fast atom bombardment tandem mass spectrometry.
Identification of fonofos metabolites in Latuca sativa, Beta vulgaris, and Triticum aestivum by packed capillary flow fast atom bombardment tandem mass spectrometry.
Journal of agricultural and food chemistry (2002-03-21)
Bruce C Onisko, Doug R Tambling, Greg W Gorder, David G Diaz, John L Ericson, Mike P Prisbylla, Chuck J Spillner
ABSTRACT
The metabolism of fonofos, a thiophosphonate insecticide, was investigated in mature lettuce (Latuca sativa), beet (Beta vulgaris), and wheat (Triticum aestivum). Six new metabolites were identified by LC-MS and LC-MS-MS analysis using fast atom bombardment (FAB) and packed capillary LC columns with application of the on-column focusing technique. These methods provided the sensitivity required to identify unknown metabolites that were present in the mature plants at only 20-230 ppb. Structural elucidation was facilitated by use of fonofos labeled with both carbon-14 and carbon-13 in the phenyl ring. In all three plants fonofos was converted to a glucose conjugate of thiophenoxylactic acid. Oxidation of the glucose conjugate gave isomeric sulfoxides in all species examined. Thiophenoxylactic acid was found esterified to malonic acid in lettuce. In beets, S-phenylcysteine was found as its malonic acid amide. A second metabolite unique to beets was N-(malonyl)-[2[(ethoxyethylphosphinothionyl)oxy]phenyl]cysteine. This novel structure was confirmed by synthesis.